Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183355.3 | NP_899198.1 | 6 - 7 | 1666 | 85 | |
GenBank mRNA | AF020196.1 | - | 6 - 7 | 1357 | 85 | |
BC058390.1 | - | 6 - 7 | 1394 | 5701148 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |