Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700019944.mm.1 |
NM_001161366.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019944.mm.1 |
NM_001161366.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019944.mm.1 |
NM_001161367.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019944.mm.1 |
NM_001161367.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019944.mm.1 |
NM_198418.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019944.mm.1 |
NM_198418.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019944.mm.1 |
TC1700002131.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019945.mm.1 |
NM_001161366.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019945.mm.1 |
NM_001161366.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019945.mm.1 |
NM_001161367.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019945.mm.1 |
NM_001161367.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019945.mm.1 |
NM_198418.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019945.mm.1 |
NM_198418.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019945.mm.1 |
TC1700002131.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019947.mm.1 |
NM_001161366.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019947.mm.1 |
NM_001161366.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019947.mm.1 |
NM_001161367.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019947.mm.1 |
NM_001161367.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019947.mm.1 |
NM_198418.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019947.mm.1 |
NM_198418.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019947.mm.1 |
TC1700002131.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019943.mm.1 |
NM_001161366.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019943.mm.1 |
NM_001161366.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019943.mm.1 |
NM_001161367.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019943.mm.1 |
NM_001161367.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019943.mm.1 |
NM_198418.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019943.mm.1 |
NM_198418.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019943.mm.1 |
TC1700002131.mm.1 |
MTA TC |