Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198858.1 | NP_942155.1 | 1 - 2 | 49 | 56 | |
GenBank mRNA | AB125661.1 | - | 1 - 2 | 49 | 56 | |
BC120763.1 | - | 1 - 2 | 41 | 40129686 | 56 | |
BC120765.1 | - | 1 - 2 | 41 | 40129688 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |