Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK005225.1 | - | 6 | 1334 | 150 | |
AK011963.1 | - | 1255 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |