Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500001162.mm.1 |
NM_001081299.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001162.mm.1 |
NM_001081299.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001162.mm.1 |
TC1500000142.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500001161.mm.1 |
NM_001081299.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001161.mm.1 |
NM_001081299.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001161.mm.1 |
TC1500000142.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500001160.mm.1 |
NM_001081299.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001160.mm.1 |
NM_001081299.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001160.mm.1 |
TC1500000142.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500001163.mm.1 |
NM_001081299.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001163.mm.1 |
NM_001081299.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001163.mm.1 |
TC1500000142.mm.1 |
MTA TC |