Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cramp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020608.3 | NP_065633.2 | 12 - 13 | 2747 | 73 | |
GenBank mRNA | AK089393.1 | - | 12 - 13 | 3007 | 73 | |
AK129356.1 | - | 3 - 4 | 923 | 73 | ||
AK147411.1 | - | 12 - 13 | 2748 | 73 | ||
AK147449.1 | - | 12 - 13 | 2748 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |