Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177052.3 | NP_796026.2 | 20 - 21 | 2270 | 104 | |
GenBank mRNA | AK084082.1 | - | 5 - 6 | 2071 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |