Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahnak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009643.2 | NP_033773.1 | 5 | 9802 | 71 | |
GenBank mRNA | BC006892.1 | - | 1 | 2359 | 71 | |
BC039931.1 | - | 1 | 2370 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |