Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A530053M12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK080055.1 | - | 1 - 2 | 79 | 74 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | PH domain and leucine rich repeat protein phosphatase 1 | Phlpp1 |
AI836256, Phlpp, Plekhe1, SCOP, mKIAA0606 |
-5.9kb | Anti-sense | Chr. 1 - 106171869 - 106394245 on NCBI Build GRCm38 |
Downstream | B cell leukemia/lymphoma 2 | Bcl2 |
AW986256, Bcl-2, C430015F12Rik, D630044D05Rik, D830018M01Rik |
136.2kb | Sense | Chr. 1 - 106538178 - 106714290 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons |