Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psma3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK169577.1 | - | 2 - 3 | 154 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |