Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK002658.1 | - | 1 - 2 | 87 | 86 | |
AK007951.1 | - | 66 | 86 | |||
AK034967.1 | - | 1 - 2 | 67 | 86 | ||
AY318751.1 | - | 74 | 86 | |||
BC061087.1 | - | 86 | 6589013 | 86 | ||
BC099551.1 | - | 58 | 30252145 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |