Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900023020.mm.1 |
NM_001198933.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023020.mm.1 |
NM_001198933.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023020.mm.1 |
NM_008615.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023020.mm.1 |
NM_008615.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023020.mm.1 |
TC0900002780.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023022.mm.1 |
NM_001198933.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023022.mm.1 |
NM_001198933.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023022.mm.1 |
NM_008615.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023022.mm.1 |
NM_008615.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023022.mm.1 |
TC0900002780.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023026.mm.1 |
NM_008615.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023026.mm.1 |
NM_008615.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023026.mm.1 |
TC0900002780.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023024.mm.1 |
NM_001198933.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023024.mm.1 |
NM_001198933.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023024.mm.1 |
NM_008615.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023024.mm.1 |
NM_008615.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023024.mm.1 |
TC0900002780.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023021.mm.1 |
NM_001198933.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023021.mm.1 |
NM_001198933.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023021.mm.1 |
NM_008615.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023021.mm.1 |
NM_008615.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023021.mm.1 |
TC0900002780.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023023.mm.1 |
NM_001198933.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023023.mm.1 |
NM_001198933.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023023.mm.1 |
NM_008615.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023023.mm.1 |
NM_008615.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023023.mm.1 |
TC0900002780.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023019.mm.1 |
NM_001198933.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023019.mm.1 |
NM_001198933.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023019.mm.1 |
NM_008615.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023019.mm.1 |
NM_008615.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023019.mm.1 |
TC0900002780.mm.1 |
MTA TC |