Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600006360.mm.1 |
NM_024218.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006360.mm.1 |
NM_024218.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006360.mm.1 |
TC1600000738.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600006353.mm.1 |
NM_024218.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006353.mm.1 |
NM_024218.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006353.mm.1 |
TC1600000738.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600006352.mm.1 |
NM_024218.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006352.mm.1 |
NM_024218.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006352.mm.1 |
TC1600000738.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600006361.mm.1 |
NM_024218.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006361.mm.1 |
NM_024218.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006361.mm.1 |
TC1600000738.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600006359.mm.1 |
NM_024218.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006359.mm.1 |
NM_024218.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006359.mm.1 |
TC1600000738.mm.1 |
MTA TC |