Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130868.2 | NP_001124340.1 | 1 | 95 | 62 | |
NM_001286384.1 | NP_001273313.1 | 1 | 95 | 62 | ||
NM_053092.3 | NP_444322.1 | 1 | 95 | 62 | ||
GenBank mRNA | BC036289.1 | - | 1 | 64 | 3157706 | 62 |
BY792549.1 | - | 1 | 46 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |