Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800014935.mm.1 |
NM_001042484.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014935.mm.1 |
NM_001042484.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014935.mm.1 |
NM_020585.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014935.mm.1 |
NM_020585.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014935.mm.1 |
TC0800001885.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800014936.mm.1 |
NM_020585.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014936.mm.1 |
NM_020585.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014936.mm.1 |
TC0800001885.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800014937.mm.1 |
NM_020585.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014937.mm.1 |
NM_020585.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014937.mm.1 |
TC0800001885.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800014929.mm.1 |
NM_001042484.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014929.mm.1 |
NM_001042484.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014929.mm.1 |
NM_020585.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014929.mm.1 |
NM_020585.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014929.mm.1 |
TC0800001885.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800014934.mm.1 |
NM_001042484.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014934.mm.1 |
NM_001042484.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014934.mm.1 |
NM_020585.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014934.mm.1 |
NM_020585.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014934.mm.1 |
TC0800001885.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800014932.mm.1 |
NM_001042484.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014932.mm.1 |
NM_001042484.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014932.mm.1 |
NM_020585.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014932.mm.1 |
NM_020585.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014932.mm.1 |
TC0800001885.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800014927.mm.1 |
NM_001042484.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014927.mm.1 |
NM_001042484.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014927.mm.1 |
NM_020585.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014927.mm.1 |
NM_020585.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014927.mm.1 |
TC0800001885.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800014928.mm.1 |
NM_001042484.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014928.mm.1 |
NM_001042484.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014928.mm.1 |
NM_020585.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014928.mm.1 |
NM_020585.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800014928.mm.1 |
TC0800001885.mm.1 |
MTA TC |