Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | spermatogenesis associated 17 | Spata17 |
1700065F16Rik, 4930504I07Rik, 4930513F16Rik, MSRG-11 |
-24.2kb | Anti-sense | Chr. 1 - 187009614 - 187215462 on NCBI Build GRCm38 |
Downstream |
predicted gene, 37896 |
Gm37896 | A930038G18 | 207.3kb | Sense | Chr. 1 - 187447697 - 187462781 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |