Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013762.2 | NP_038790.2 | 5 - 6 | 769 | 100 | |
GenBank mRNA | AK088993.1 | - | 5 - 6 | 716 | 100 | |
AK144518.1 | - | 5 - 6 | 716 | 100 | ||
AK144816.1 | - | 5 - 6 | 716 | 100 | ||
AK146502.1 | - | 5 - 6 | 716 | 100 | ||
AK149494.1 | - | 5 - 6 | 716 | 100 | ||
AK151057.1 | - | 5 - 6 | 716 | 100 | ||
AK151084.1 | - | 5 - 6 | 716 | 100 | ||
AK151142.1 | - | 5 - 6 | 716 | 100 | ||
AK151426.1 | - | 5 - 6 | 716 | 100 | ||
AK151596.1 | - | 5 - 6 | 716 | 100 | ||
AK151740.1 | - | 5 - 6 | 716 | 100 | ||
AK166433.1 | - | 5 - 6 | 712 | 100 | ||
AK166825.1 | - | 5 - 6 | 716 | 100 | ||
AK167787.1 | - | 5 - 6 | 716 | 100 | ||
AK167947.1 | - | 5 - 6 | 716 | 100 | ||
AK168040.1 | - | 5 - 6 | 716 | 100 | ||
AK168059.1 | - | 5 - 6 | 715 | 100 | ||
AK168126.1 | - | 5 - 6 | 716 | 100 | ||
AK168128.1 | - | 5 - 6 | 716 | 100 | ||
AK168224.1 | - | 5 - 6 | 716 | 100 | ||
AK168744.1 | - | 5 - 6 | 714 | 100 | ||
AK169060.1 | - | 5 - 6 | 716 | 100 | ||
AK169235.1 | - | 5 - 6 | 716 | 100 | ||
AK169321.1 | - | 5 - 6 | 716 | 100 | ||
AK170819.1 | - | 5 - 6 | 716 | 100 | ||
AK172280.1 | - | 5 - 6 | 716 | 100 | ||
AK190439.1 | - | 1 - 2 | 92 | 100 | ||
AK197531.1 | - | 1 - 2 | 92 | 100 | ||
AK215968.1 | - | 1 - 2 | 92 | 100 | ||
BC009655.1 | - | 5 - 6 | 712 | 3485069 | 100 | |
BC083134.1 | - | 4 - 5 | 698 | 6504713 | 100 | |
BC094059.1 | - | 4 - 5 | 697 | 3676103 | 100 | |
BC098497.1 | - | 3 - 4 | 1274 | 100 | ||
BC158039.1 | - | 5 - 6 | 716 | 9007342 | 100 | |
U89417.1 | - | 1 - 2 | 144 | 100 | ||
Y00225.1 | - | 4 - 5 | 698 | 100 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AK219709.1 | - | 92 | Chr. - - [-] on Build | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |