Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205385.1 | NP_001192314.1 | 6 - 7 | 634 | 149 | |
NM_001205386.1 | NP_001192315.1 | 6 - 7 | 631 | 149 | ||
NM_023735.2 | NP_076224.1 | 5 - 6 | 826 | 149 | ||
GenBank mRNA | AK004554.1 | - | 5 - 6 | 640 | 149 | |
AK083343.1 | - | 5 - 6 | 644 | 149 | ||
AK145536.1 | - | 5 - 6 | 646 | 149 | ||
AK150487.1 | - | 5 - 6 | 528 | 149 | ||
AK150589.1 | - | 5 - 6 | 604 | 149 | ||
AK150991.1 | - | 5 - 6 | 584 | 149 | ||
AK152865.1 | - | 5 - 6 | 621 | 149 | ||
AK153079.1 | - | 5 - 6 | 647 | 149 | ||
AK153150.1 | - | 5 - 6 | 621 | 149 | ||
AK153204.1 | - | 5 - 6 | 527 | 149 | ||
AK159135.1 | - | 5 - 6 | 613 | 149 | ||
AK159668.1 | - | 5 - 6 | 538 | 149 | ||
AK159775.1 | - | 5 - 6 | 625 | 149 | ||
AK160047.1 | - | 5 - 6 | 486 | 149 | ||
AK166529.1 | - | 6 - 7 | 578 | 149 | ||
AK166586.1 | - | 6 - 7 | 578 | 149 | ||
AK167148.1 | - | 5 - 6 | 585 | 149 | ||
AK167891.1 | - | 5 - 6 | 583 | 149 | ||
AK168568.1 | - | 5 - 6 | 513 | 149 | ||
AK168771.1 | - | 5 - 6 | 599 | 149 | ||
BC005557.1 | - | 5 - 6 | 589 | 3156748 | 149 | |
BC080806.1 | - | 6 - 7 | 592 | 5350752 | 149 | |
BC108386.1 | - | 5 - 6 | 557 | 5051125 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |