Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mprip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012027.2 | NP_036157.2 | 17 - 18 | 2759 | 110 | |
NM_201245.3 | NP_957697.1 | 17 - 18 | 2759 | 110 | ||
GenBank mRNA | AB093269.1 | - | 5 - 6 | 4584 | 110 | |
AK035435.1 | - | 11 - 12 | 2049 | 110 | ||
AK154807.1 | - | 18 - 19 | 2766 | 110 | ||
AK155630.1 | - | 17 - 18 | 2540 | 110 | ||
AK158296.1 | - | 17 - 18 | 2748 | 110 | ||
AK162530.1 | - | 15 - 16 | 2105 | 110 | ||
AK170849.1 | - | 17 - 18 | 2743 | 110 | ||
AK171089.1 | - | 17 - 18 | 2748 | 110 | ||
BC027417.1 | - | 3 - 4 | 848 | 110 | ||
BC039932.1 | - | 16 - 17 | 2283 | 110 | ||
BC049803.1 | - | 17 - 18 | 2285 | 110 | ||
BC063067.1 | - | 17 - 18 | 2722 | 5721396 | 110 | |
U73200.1 | - | 17 - 18 | 2619 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |