Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxx1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC031701.1 | - | 1 - 2 | 417 | 3594379 | 176 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |