Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004143.4 | NP_001004143.2 | 5 - 6 | 860 | 132 | |
GenBank mRNA | AK034492.1 | - | 5 - 6 | 835 | 132 | |
AK155071.1 | - | 5 - 6 | 861 | 132 | ||
AK160992.1 | - | 5 - 6 | 769 | 132 | ||
AK220368.1 | - | 5 - 6 | 821 | 132 | ||
BC052540.1 | - | 1 - 2 | 155 | 132 | ||
BC080737.1 | - | 5 - 6 | 731 | 30431735 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |