Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skint4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178786.4 | NP_848901.2 | 8 - 9 | 1351 | 148 | |
GenBank mRNA | AK028550.1 | - | 8 - 9 | 1403 | 148 | |
AK028795.1 | - | 8 - 9 | 1352 | 148 | ||
AK035748.1 | - | 8 - 9 | 1269 | 148 | ||
AK162813.1 | - | 9 - 10 | 1366 | 148 | ||
EF494897.1 | - | 8 - 9 | 1150 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |