Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700013112.mm.1 |
NM_199252.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013112.mm.1 |
NM_199252.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013112.mm.1 |
TC1700001465.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013113.mm.1 |
NM_199252.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013113.mm.1 |
NM_199252.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013113.mm.1 |
TC1700001465.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012594.mm.1 |
NM_001142539.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012594.mm.1 |
NM_001142539.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012594.mm.1 |
TC1700001400.mm.1 |
MTA TC |