Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007463.1 | NP_001007464.1 | 2 - 3 | 832 | 133 | |
NM_001290462.1 | NP_001277391.1 | 2 - 3 | 832 | 133 | ||
GenBank mRNA | AK132968.1 | - | 2 - 3 | 833 | 133 | |
AY769082.1 | - | 2 - 3 | 832 | 133 | ||
BC150868.1 | - | 2 - 3 | 874 | 9087779 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |