Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200021555.mm.1 |
NM_080455.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021555.mm.1 |
NM_080455.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021555.mm.1 |
TC0200002632.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200021550.mm.1 |
NM_080455.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021550.mm.1 |
NM_080455.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021550.mm.1 |
TC0200002632.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200021552.mm.1 |
NM_080455.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021552.mm.1 |
NM_080455.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021552.mm.1 |
TC0200002632.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200021551.mm.1 |
NM_080455.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021551.mm.1 |
NM_080455.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021551.mm.1 |
TC0200002632.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200021553.mm.1 |
NM_080455.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021553.mm.1 |
NM_080455.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021553.mm.1 |
TC0200002632.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200021554.mm.1 |
NM_080455.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021554.mm.1 |
NM_080455.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021554.mm.1 |
TC0200002632.mm.1 |
MTA TC |