Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134426.1 | NP_001127898.1 | 1 - 2 | 729 | 71 | |
NM_175565.3 | NP_780774.1 | 1 - 2 | 729 | 71 | ||
NM_175833.2 | NP_787027.1 | 1 - 2 | 729 | 71 | ||
GenBank mRNA | AB046372.1 | - | 1 - 2 | 317 | 71 | |
AF215660.1 | - | 1 - 2 | 487 | 71 | ||
AF320340.1 | - | 1 - 2 | 487 | 71 | ||
AK010956.1 | - | 1 - 2 | 621 | 71 | ||
BC052770.1 | - | 1 - 2 | 623 | 6306557 | 71 | |
BC062156.1 | - | 2 - 3 | 575 | 6507616 | 71 | |
BC096479.1 | - | 1 - 2 | 580 | 4979176 | 71 | |
L22067.1 | - | 1 - 2 | 570 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |