Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fzd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020510.2 | NP_065256.1 | 1 | 1991 | 73 | |
GenBank mRNA | AF206321.1 | - | 1 | 1945 | 73 | |
AF206322.1 | - | 1 | 1021 | 73 | ||
AF363723.1 | - | 1 | 773 | 73 | ||
AK048420.1 | - | 1 | 1968 | 73 | ||
AK135363.1 | - | 1 | 1954 | 73 | ||
BC049774.1 | - | 1 | 1808 | 6517721 | 73 | |
BC055727.1 | - | 1 | 1970 | 6411627 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Mouse Wnt Panel 1700 Inventoried Both primers and probe map within a single exon |