Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017966.2 | NP_001017966.1 | 8 - 9 | 1408 | 85 | |
GenBank mRNA | BC021415.1 | - | 8 - 9 | 1389 | 85 | |
BC031491.1 | - | 1 - 2 | 162 | 85 | ||
CD550969.1 | - | 3 - 4 | 320 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |