Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200026901.mm.1 |
NM_001033908.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026901.mm.1 |
NM_001033908.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026901.mm.1 |
NM_011513.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026901.mm.1 |
NM_011513.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026901.mm.1 |
TC0200003203.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200026902.mm.1 |
NM_001033908.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026902.mm.1 |
NM_001033908.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026902.mm.1 |
NM_011513.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026902.mm.1 |
NM_011513.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026902.mm.1 |
TC0200003203.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200026906.mm.1 |
NM_001033908.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026906.mm.1 |
NM_001033908.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026906.mm.1 |
NM_011513.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026906.mm.1 |
NM_011513.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026906.mm.1 |
TC0200003203.mm.1 |
MTA TC |