Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500031589.mm.1 |
TC0500003465.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500031589.mm.1 |
NM_001024932.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031589.mm.1 |
NM_001024932.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031591.mm.1 |
TC0500003465.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500031591.mm.1 |
NM_001024932.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031591.mm.1 |
NM_001024932.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031590.mm.1 |
TC0500003465.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500031590.mm.1 |
NM_001024932.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031590.mm.1 |
NM_001024932.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031588.mm.1 |
TC0500003465.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500031588.mm.1 |
NM_001024932.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031588.mm.1 |
NM_001024932.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031587.mm.1 |
TC0500003465.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500031587.mm.1 |
NM_001024932.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031587.mm.1 |
NM_001024932.2 |
Refseq |