Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100034159.mm.1 |
NM_001033397.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034159.mm.1 |
NM_001033397.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034159.mm.1 |
TC1100003703.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034158.mm.1 |
NM_001033397.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034158.mm.1 |
NM_001033397.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034158.mm.1 |
TC1100003703.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034161.mm.1 |
NM_001033397.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034161.mm.1 |
NM_001033397.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034161.mm.1 |
TC1100003703.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034165.mm.1 |
NM_001033397.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034165.mm.1 |
NM_001033397.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034165.mm.1 |
TC1100003703.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034160.mm.1 |
NM_001033397.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034160.mm.1 |
NM_001033397.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034160.mm.1 |
TC1100003703.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034166.mm.1 |
NM_001033397.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034166.mm.1 |
NM_001033397.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034166.mm.1 |
TC1100003703.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034162.mm.1 |
NM_001033397.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034162.mm.1 |
NM_001033397.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034162.mm.1 |
TC1100003703.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034164.mm.1 |
NM_001033397.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034164.mm.1 |
NM_001033397.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034164.mm.1 |
TC1100003703.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034163.mm.1 |
NM_001033397.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034163.mm.1 |
NM_001033397.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034163.mm.1 |
TC1100003703.mm.1 |
MTA TC |