Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033408.4 | NP_001028580.1 | 8 - 9 | 2174 | 77 | |
GenBank mRNA | AK133355.1 | - | 8 - 9 | 2175 | 77 | |
BC150950.1 | - | 8 - 9 | 2136 | 9087861 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |