Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200029705.mm.1 |
NM_001289521.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029705.mm.1 |
NM_015753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029705.mm.1 |
TC0200003473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200029708.mm.1 |
NM_001289521.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029708.mm.1 |
NM_015753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029708.mm.1 |
TC0200003473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200029707.mm.1 |
NM_001289521.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029707.mm.1 |
NM_015753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029707.mm.1 |
TC0200003473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200029710.mm.1 |
NM_001289521.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029710.mm.1 |
NM_015753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029710.mm.1 |
TC0200003473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200029709.mm.1 |
NM_001289521.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029709.mm.1 |
NM_015753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029709.mm.1 |
TC0200003473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200029704.mm.1 |
NM_001289521.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029704.mm.1 |
NM_015753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029704.mm.1 |
TC0200003473.mm.1 |
MTA TC |