Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnaset2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026611.2 | NP_080887.1 | 4 - 5 | 413 | 68 | |
GenBank mRNA | AK029149.1 | - | 367 | 68 | ||
AK084454.1 | - | 373 | 68 | |||
BC100330.1 | - | 4 - 5 | 338 | 30527517 | 68 |
Gene Symbol: Rnaset2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083938.2 | NP_001077407.1 | 4 - 5 | 369 | 68 | |
GenBank mRNA | AK014754.1 | - | 1 - 2 | 212 | 68 | |
AK018722.1 | - | 4 - 5 | 369 | 68 | ||
AK019234.1 | - | 4 - 5 | 361 | 68 | ||
BC031496.1 | - | 4 - 5 | 349 | 3156549 | 68 | |
BC089534.1 | - | 4 - 5 | 678 | 6703307 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |