Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npbwr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010342.1 | NP_034472.1 | 1 | 3312 | 118 | |
GenBank mRNA | AK138357.1 | - | 1 | 3317 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon |