Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037927.1 | NP_001033016.1 | 14 - 15 | 1636 | 65 | |
GenBank mRNA | AB205021.1 | - | 14 - 15 | 1636 | 65 | |
BC150920.1 | - | 13 - 14 | 1606 | 9087831 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |