Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kncn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039124.3 | NP_001034213.1 | 2 - 3 | 560 | 73 | |
GenBank mRNA | BC132034.1 | - | 2 - 3 | 507 | 40130311 | 73 |
BC132036.1 | - | 2 - 3 | 507 | 40130313 | 73 | |
DQ235452.1 | - | 2 - 3 | 560 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |