Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016897.3 | NP_058593.2 | 5 - 6 | 550 | 91 | |
GenBank mRNA | AB021122.1 | - | 5 - 6 | 530 | 91 | |
AK013976.1 | - | 5 - 6 | 496 | 91 | ||
AK146516.1 | - | 5 - 6 | 516 | 91 | ||
BC037022.1 | - | 6 - 7 | 527 | 5366968 | 91 | |
BC062963.1 | - | 5 - 6 | 486 | 5692352 | 91 | |
BC069903.1 | - | 5 - 6 | 520 | 5290493 | 91 | |
BC096669.1 | - | 5 - 6 | 519 | 5056242 | 91 | |
BC132019.1 | - | 5 - 6 | 499 | 40130296 | 91 | |
BC132319.1 | - | 5 - 6 | 499 | 40130596 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |