Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020252.3 | NP_064648.3 | 14 - 15 | 3957 | 77 | |
NM_177284.2 | NP_796258.2 | 12 - 13 | 3921 | 77 | ||
GenBank mRNA | AB093249.2 | - | 14 - 15 | 3957 | 77 | |
AK133271.1 | - | 9 - 10 | 2548 | 77 | ||
AK137407.1 | - | 13 - 14 | 3941 | 77 | ||
BC079893.1 | - | 12 - 13 | 3852 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |