Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Susd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025491.3 | NP_079767.2 | 3 - 4 | 459 | 86 | |
NM_028340.1 | NP_082616.1 | 3 - 4 | 628 | 86 | ||
GenBank mRNA | AK003860.1 | - | 3 - 4 | 459 | 86 | |
AK006068.1 | - | 3 - 4 | 449 | 86 | ||
BC045525.1 | - | 3 - 4 | 628 | 6468669 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |