Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf3c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033194.2 | NP_001028366.1 | 7 - 8 | 1081 | 142 | |
GenBank mRNA | AK145040.1 | - | 1083 | 142 | ||
AK160997.1 | - | 7 - 8 | 1069 | 142 | ||
AK165830.1 | - | 7 - 8 | 1082 | 142 | ||
BC141157.1 | - | 7 - 8 | 1071 | 9055769 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |