Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007559.4 | NP_031585.2 | 5 - 6 | 1128 | 103 | |
GenBank mRNA | AK038590.1 | - | 350 | 103 | ||
AK082895.1 | - | 5 - 6 | 1129 | 103 | ||
BC137890.1 | - | 5 - 6 | 998 | 8860909 | 103 | |
U39545.1 | - | 5 - 6 | 1128 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |