Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009757.4 | NP_033887.1 | 2 | 769 | 111 | |
GenBank mRNA | AF082348.1 | - | 2 | 652 | 111 | |
AJ132406.1 | - | 2 | 409 | 111 | ||
AK028735.1 | - | 2 | 455 | 111 | ||
AK135757.1 | - | 2 | 676 | 111 | ||
AK135882.1 | - | 2 | 772 | 111 | ||
AK136127.1 | - | 2 | 163 | 111 | ||
AK136129.1 | - | 2 | 163 | 111 | ||
AK139773.1 | - | 2 | 770 | 111 | ||
AK139847.1 | - | 2 | 771 | 111 | ||
AK139936.1 | - | 2 | 725 | 111 | ||
BC055363.1 | - | 2 | 766 | 30051425 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon |