Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145741.2 | NP_665684.2 | 2 | 1231 | 102 | |
GenBank mRNA | AK042684.1 | - | 1222 | 102 | ||
BC022669.1 | - | 1 | 541 | 102 | ||
BC058358.1 | - | 2 | 1231 | 6828166 | 102 | |
L42114.1 | - | 2 | 1047 | 102 | ||
S82648.1 | - | 2 | 1047 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon |