Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10447518 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10447519 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10447517 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1700012230.mm.1 |
NR_003519.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012230.mm.1 |
TC1700001348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012234.mm.1 |
NR_003519.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012234.mm.1 |
TC1700001348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012233.mm.1 |
NR_003519.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012233.mm.1 |
TC1700001348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012245.mm.1 |
NR_003519.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012245.mm.1 |
TC1700001348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012253.mm.1 |
NR_003519.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012253.mm.1 |
TC1700001348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012255.mm.1 |
NR_003519.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012255.mm.1 |
TC1700001348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012229.mm.1 |
NR_003519.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012229.mm.1 |
TC1700001348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012257.mm.1 |
NR_003519.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012257.mm.1 |
TC1700001348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012232.mm.1 |
NR_003519.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012232.mm.1 |
TC1700001348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012236.mm.1 |
NR_003519.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012236.mm.1 |
TC1700001348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012237.mm.1 |
NR_003519.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012237.mm.1 |
TC1700001348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012239.mm.1 |
NR_003519.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012239.mm.1 |
TC1700001348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012254.mm.1 |
NR_003519.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012254.mm.1 |
TC1700001348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012246.mm.1 |
NR_003519.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012246.mm.1 |
TC1700001348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012243.mm.1 |
NR_003519.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012243.mm.1 |
TC1700001348.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1439070_PM_x_at |
- |
- |
Mouse Expression Set 430 |
1439070_x_at |
- |
- |
Mouse Expression Set 430 |
1454566_at |
- |
- |
Mouse Expression Set 430 |
1454566_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1439070_PM_x_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1439070_x_at |
- |
- |