Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026130.1 | NP_080406.1 | 4 - 5 | 612 | 146 | |
GenBank mRNA | AK004967.2 | - | 4 - 5 | 612 | 146 | |
AK038017.1 | - | 4 - 5 | 600 | 146 | ||
BC021839.1 | - | 4 - 5 | 595 | 4207633 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |