Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100015722.mm.1 |
NM_001195662.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015722.mm.1 |
NM_001195662.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015722.mm.1 |
TC0100001904.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015731.mm.1 |
NM_001195662.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015731.mm.1 |
NM_001195662.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015731.mm.1 |
TC0100001904.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015723.mm.1 |
NM_001195662.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015723.mm.1 |
NM_001195662.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015723.mm.1 |
TC0100001904.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015729.mm.1 |
NM_011283.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015729.mm.1 |
NM_011283.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015729.mm.1 |
TC0100001904.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015728.mm.1 |
NM_001195662.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015728.mm.1 |
NM_001195662.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015728.mm.1 |
NM_011283.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015728.mm.1 |
NM_011283.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015728.mm.1 |
TC0100001904.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015727.mm.1 |
NM_001195662.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015727.mm.1 |
NM_001195662.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015727.mm.1 |
NM_011283.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015727.mm.1 |
NM_011283.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015727.mm.1 |
TC0100001904.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015730.mm.1 |
NM_011283.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015730.mm.1 |
NM_011283.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015730.mm.1 |
TC0100001904.mm.1 |
MTA TC |