Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500013783.mm.1 |
TC0500001526.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500013783.mm.1 |
NM_001199059.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500013783.mm.1 |
NM_001199059.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500013786.mm.1 |
TC0500001526.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500013786.mm.1 |
NM_019983.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500013786.mm.1 |
NM_019983.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500013785.mm.1 |
TC0500001526.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500013785.mm.1 |
NM_019983.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500013785.mm.1 |
NM_019983.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500013788.mm.1 |
TC0500001526.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500013788.mm.1 |
NM_001199059.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500013788.mm.1 |
NM_001199059.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500013788.mm.1 |
NM_019983.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500013788.mm.1 |
NM_019983.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500013787.mm.1 |
TC0500001526.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500013787.mm.1 |
NM_001199059.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500013787.mm.1 |
NM_001199059.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500013787.mm.1 |
NM_019983.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500013787.mm.1 |
NM_019983.2 |
Refseq |