Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130868.2 | NP_001124340.1 | 13 - 14 | 1815 | 99 | |
NM_001286384.1 | NP_001273313.1 | 14 - 15 | 1873 | 99 | ||
NM_053092.3 | NP_444322.1 | 12 - 13 | 1635 | 99 | ||
GenBank mRNA | AK089053.1 | - | 12 - 13 | 1560 | 99 | |
AK090203.1 | - | 12 - 13 | 1560 | 99 | ||
AK153236.1 | - | 12 - 13 | 1558 | 99 | ||
AK167693.1 | - | 12 - 13 | 1560 | 99 | ||
BC027356.1 | - | 14 - 15 | 1785 | 4950333 | 99 | |
BC035324.1 | - | 13 - 14 | 1727 | 2811286 | 99 | |
BC036289.1 | - | 12 - 13 | 1604 | 3157706 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |