Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcamr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144960.2 | NP_659209.2 | 1 - 2 | 92 | 87 | |
GenBank mRNA | AB048834.1 | - | 1 - 2 | 104 | 87 | |
BC110550.1 | - | 1 - 2 | 95 | 40047153 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |